accessibility menu, dialog, popup

UserName

CAS RN 22924-15-8

CH3OO

CAS RN 22924-15-8

IUPAC Name: 3-ethoxybenzaldehyde
Synonyms: benzaldehyde, 3-ethoxy-
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: QZMGMXBYJZVAJN-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(C=O)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)