missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. E11335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11335G 5 g
E113325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11335G Supplier TCI America Supplier No. E11335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22924-15-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00016606
InChI Key QZMGMXBYJZVAJN-UHFFFAOYSA-N
Synonym m-ethoxybenzaldehyde, benzaldehyde, 3-ethoxy, 3-ethoxy-benzaldehyde, benzaldehyde, m-ethoxy, 3-ethoxy benzaldehyde, 3-ethoxybenzaldehdye, pubchem13132, acmc-209fzo, intermediates-zcf02615, 3-ethoxybenzaldehyde
PubChem CID 89908
IUPAC Name 3-ethoxybenzaldehyde
SMILES CCOC1=CC=CC(=C1)C=O

Specifications

Color Yellow
Boiling Point 245°C
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethoxybenzaldehyde
EINECSNumber (3)-2662
TSCA No