accessibility menu, dialog, popup

UserName

OHNOOClONNClClCl

CAS RN 22832-87-7

IUPAC Name: 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole; nitric acid
Synonyms: miconazole nitrate
Molecular Weight (g/mol): 479.14
Molecular Formula: C18H15Cl4N3O4
InChi Key: MCCACAIVAXEFAL-UHFFFAOYNA-N
SMILES: O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)
  • (2)
Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)