missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Miconazole nitrate, 98+%

Catalog No. AAJ6079614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6079614 25 g
AAJ6079606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6079614 Supplier Thermo Scientific Chemicals Supplier No. J6079614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibits cytochrome P450-dependent 14a-demethylase | CAS: 22832-87-7 | C18H15Cl4N3O4 | 479.14 g/mol

Inhibits cytochrome P450-dependent 14a-demethylase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22832-87-7
Molecular Formula C18H15Cl4N3O4
Molecular Weight (g/mol) 479.14
MDL Number MFCD00058161
InChI Key MCCACAIVAXEFAL-UHFFFAOYNA-N
Synonym miconazole nitrate, albistat, andergin, aflorix, conofite, florid, micatin, gyno-monistat, epi-monistat, gyno-daktar
PubChem CID 68553
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole; nitric acid
SMILES O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1

Specifications

Melting Point 170°C to 185°C
Color White
Quantity 25 g
Merck Index 14,6178
Solubility Information Slightly soluble in water; Soluble in propylene glycol or pyridine.
Formula Weight 479.15
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material (+/-)-Miconazole nitrate
Hazard Category H302-H317
Hazard Statement GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
EINECSNumber 245-256-6
RTECSNumber NI4771000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only