CAS RN 22734-04-9
IUPAC Name:
2-(cyclopent-1-en-1-yl)acetonitrile
Synonyms:
2-(cyclopent-1-en-1-yl)acetonitrile
Molecular Weight (g/mol):
107.16
Molecular Formula:
C7H9N
InChi Key:
LEWVRAMNXUWSFL-UHFFFAOYSA-N
SMILES:
N#CCC1=CCCC1
1
–
1
of
1
results
1
1-Cyclopentenylacetonitrile 90.0+%, TCI America™
CAS: 22734-04-9 Molecular Formula: C7H9N Molecular Weight (g/mol): 107.16 MDL Number: MFCD00001398 InChI Key: LEWVRAMNXUWSFL-UHFFFAOYSA-N Synonym: 1-cyclopentenylacetonitrile,1-cyanomethyl cyclopentene,2-cyclopent-1-en-1-yl acetonitrile,cyclopent-1-ene-1-acetonitrile,1-cyclopentylacetonitrile,1-cyclopenteneacetonitrile,1-cyclopentene-1-acetonitrile,1-cyclopentenylacetronitrile,2-cyclopenten-1-yl acetonitrile PubChem CID: 89811 IUPAC Name: 2-(cyclopent-1-en-1-yl)acetonitrile SMILES: N#CCC1=CCCC1