missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentenylacetonitrile 90.0+%, TCI America™

Catalog No. C125310ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C125310ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C125310ML Supplier TCI America Supplier No. C125310ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22734-04-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00001398
InChI Key LEWVRAMNXUWSFL-UHFFFAOYSA-N
Synonym 1-cyclopentenylacetonitrile, 1-cyanomethyl cyclopentene, 2-cyclopent-1-en-1-yl acetonitrile, cyclopent-1-ene-1-acetonitrile, 1-cyclopentylacetonitrile, 1-cyclopenteneacetonitrile, 1-cyclopentene-1-acetonitrile, 1-cyclopentenylacetronitrile, 2-cyclopenten-1-yl acetonitrile
PubChem CID 89811
IUPAC Name 2-(cyclopent-1-en-1-yl)acetonitrile
SMILES N#CCC1=CCCC1

Specifications

Color Colorless
Boiling Point 192°C
Quantity 10 mL
UN Number 3276
Formula Weight 107.16
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Cyclopentenylacetonitrile