accessibility menu, dialog, popup

UserName

CAS RN 226721-96-6

H3COOOH2OH2ONa

CAS RN 226721-96-6

IUPAC Name: sodium 2-{4-[(2-oxocyclopentyl)methyl]phenyl}propanoate dihydrate
Synonyms: sodium 2-{4-[(2-oxocyclopentyl)methyl]phenyl}propanoate dihydrate
Molecular Weight (g/mol): 304.32
Molecular Formula: C15H21NaO5
InChi Key: BAZQYVYVKYOAGO-UHFFFAOYNA-M
SMILES: O.O.[Na+].CC(C([O-])=O)C1=CC=C(CC2CCCC2=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results