missing translation for 'onlineSavingsMsg'
Learn More

Loxoprofen Sodium Salt Dihydrate 98.0+%, TCI America™

Catalog No. L02525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02525G 5 g
L025225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02525G Supplier TCI America Supplier No. L02525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 226721-96-6
Molecular Formula C15H21NaO5
Molecular Weight (g/mol) 304.318
MDL Number MFCD01745788
InChI Key BAZQYVYVKYOAGO-UHFFFAOYSA-M
Synonym Sodium 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionate, 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionic Acid Sodium Salt
PubChem CID 23674745
ChEBI CHEBI:31786
IUPAC Name sodium;2-[4-[(2-oxocyclopentyl)methyl]phenyl]propanoate;dihydrate
SMILES CC(C1=CC=C(C=C1)CC2CCCC2=O)C(=O)[O-].O.O.[Na+]

Specifications

Melting Point 198°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 268.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Loxoprofen Sodium Salt Dihydrate
RTECSNumber CY1678845
TSCA No