accessibility menu, dialog, popup

UserName

H2CNZnBr

CAS RN 226570-68-9

IUPAC Name: zinc(2+) 4-cyanobutan-1-ide bromide
Synonyms: zinc(2+) 4-cyanobutan-1-ide bromide
Molecular Weight (g/mol): 227.41
Molecular Formula: C5H8BrNZn
InChi Key: WGCKXFGNNBONBG-UHFFFAOYSA-M
SMILES: [Zn++].[Br-].[CH2-]CCCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)