missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobutylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58897AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58897AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58897AD Supplier Thermo Scientific Chemicals Supplier No. H58897AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 226570-68-9 | C5H9BrNZn- | 228.418 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary intermediate.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids a

Chemical Identifiers

CAS 226570-68-9
Molecular Formula C5H9BrNZn-
Molecular Weight (g/mol) 228.418
MDL Number MFCD01316996
InChI Key VKWWIKWQDKNXQC-UHFFFAOYSA-M
Synonym 4-cyanobutylzinc bromide 0.5m in thf
PubChem CID 131668629
IUPAC Name pentanenitrile;zinc;bromide
SMILES CCCCC#N.[Zn].[Br-]

Specifications

Density 0.971
Linear Formula N≡C(CH2)4ZnBr
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Solubility Information Reacts with water.
Formula Weight 227.41
Chemical Name or Material 4-Cyanobutylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H302-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only