accessibility menu, dialog, popup

UserName

CH3H2COOFFFFFFFFFFFF

CAS RN 2261-99-6

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl 2-methylprop-2-enoate
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 400.16
Molecular Formula: C11H8F12O2
InChi Key: YJKHMSPWWGBKTN-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results