accessibility menu, dialog, popup

UserName

CAS RN 22385-77-9

IUPAC Name: 1-bromo-3,5-di-tert-butylbenzene
Synonyms: 1-bromo-3,5-di-tert-butylbenzene
Molecular Weight (g/mol): 269.23
Molecular Formula: C14H21Br
InChi Key: BUOWTUULDKULFI-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC(Br)=C1)C(C)(C)C
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)