missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-di-tert-butylbenzene 98.0+%, TCI America™

Catalog No. B35475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35475G 5 g
B354725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35475G Supplier TCI America Supplier No. B35475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22385-77-9
Molecular Formula C14H21Br
Molecular Weight (g/mol) 269.226
MDL Number MFCD00796945
InChI Key BUOWTUULDKULFI-UHFFFAOYSA-N
Synonym 1-bromo-3,5-di-tert-butylbenzene, 3,5-di-tert-butylbromobenzene, 3,5-di-t-butylbromobenzene, 1-bromo-3,5-di-t-butylbenzene, 1-bromo-3,5-ditert-butyl benzene, benzene, 1-bromo-3,5-bis 1,1-dimethylethyl, 1,5-bis tert-butyl-3-bromobenzene, acmc-1cpbo, g00018-watson-int, 3,5-di-t-butyl-bromobenzene
PubChem CID 620136
IUPAC Name 1-bromo-3,5-ditert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC(=C1)Br)C(C)(C)C

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 158°C
Quantity 5 g
Formula Weight 269.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-3,5-di-tert-butylbenzene