accessibility menu, dialog, popup

UserName

CAS RN 22306-37-2

BiCH3OOCH3OOCH3OO

CAS RN 22306-37-2

IUPAC Name: bismuth(3+) triacetate
Synonyms: bismuth(3+) triacetate
Molecular Weight (g/mol): 386.11
Molecular Formula: C6H9BiO6
InChi Key: WKLWZEWIYUTZNJ-UHFFFAOYSA-K
SMILES: [Bi+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)