missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) acetate, 99.999% (metals basis)

Catalog No. AA1184618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1184618 50 g
AA1184609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1184618 Supplier Thermo Scientific Chemicals Supplier No. 01184618

CAS: 22306-37-2 | C6H9BiO6 | 386.112 g/mol

It is used in biology, printing, in photographic films, textiles, and also in proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22306-37-2
Molecular Formula C6H9BiO6
Molecular Weight (g/mol) 386.112
MDL Number MFCD00015629
InChI Key WKLWZEWIYUTZNJ-UHFFFAOYSA-K
Synonym bismuth acetate, bismuth triacetate, unii-8aja86y692, acetic acid, bismuth 3+ salt, acmc-20allk, bismuth iii ; acetate, bismuth 3+ triacetate, trisacetic acid bismuth salt, bismuth 3+ ion tris acetate ion, bismuth iii acetate 50g
PubChem CID 31132
IUPAC Name bismuth;triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Bi+3]

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystals
Quantity 50 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in water.
Formula Weight 386.12
Sensitivity Hygroscopic
Chemical Name or Material Bismuth(III) acetate
EINECSNumber 244-904-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only