accessibility menu, dialog, popup

UserName

CH3NClNOO

CAS RN 22280-60-0

IUPAC Name: 6-chloro-2-methyl-3-nitropyridine
Synonyms: 6-chloro-2-methyl-3-nitropyridine
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: GHSRMSJVYMITDX-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=CC=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)