missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-methyl-3-nitropyridine 98.0+%, TCI America™

Catalog No. C24345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24345G 5 g
C24341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24345G Supplier TCI America Supplier No. C24345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22280-60-0 | C6H5ClN2O2 | 172.568 g/mol

Chemical Identifiers

CAS 22280-60-0
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00234160
InChI Key GHSRMSJVYMITDX-UHFFFAOYSA-N
Synonym 2-chloro-5-nitro-6-methylpyridine, 6-chloro-3-nitro-2-picoline, 2-chloro-5-nitro-6-picoline, 6-chloro-2-methyl-3-nitro-pyridine, 6-chloro-3-nitro-2-methylpyridine, pyridine, 6-chloro-2-methyl-3-nitro, 2-chloro-6-methyl-5-nitropyridine, 2-chloro-5-nitro-6-picoline 2-chloro-6-methyl-5-nitropyridine, pubchem6225, acmc-1cg7s
PubChem CID 253123
IUPAC Name 6-chloro-2-methyl-3-nitropyridine
SMILES CC1=C(C=CC(=N1)Cl)[N+](=O)[O-]

Specifications

Melting Point 55°C
Quantity 5 g
Formula Weight 172.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-2-methyl-3-nitropyridine