accessibility menu, dialog, popup

UserName

CAS RN 22252-43-3

IUPAC Name: 7-amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms: 7-amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molecular Weight (g/mol): 214.24
Molecular Formula: C8H10N2O3S
InChi Key: NVIAYEIXYQCDAN-UHFFFAOYNA-N
SMILES: CC1=C(N2C(SC1)C(N)C2=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results