missing translation for 'onlineSavingsMsg'
Learn More

7-Aminodesacetoxycephalosporanic Acid 98.0+%, TCI America™

Catalog No. A20755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20755G 5 g
A207525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20755G Supplier TCI America Supplier No. A20755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22252-43-3
Molecular Formula C8H10N2O3S
Molecular Weight (g/mol) 214.239
MDL Number MFCD00151456
InChI Key NVIAYEIXYQCDAN-CLZZGJSISA-N
Synonym 7-aminodesacetoxycephalosporanic acid, 6r,7r-7-amino-3-methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, 7-aminodeacetoxycephalosporanic acid, 7-adca, unii-anm3msm8tn, anm3msm8tn, 7beta-amino-3-methyl-3-cephem-4-carboxylic acid, 7beta-amino-3-methylceph-3-em-4-carboxylic acid, 7-amino desacetoxy cephalosporanic acid, 7beta-aminodeacetoxycephalosporanic acid
PubChem CID 33498
ChEBI CHEBI:64984
IUPAC Name (6R,7R)-7-amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC1=C(N2C(C(C2=O)N)SC1)C(=O)O

Specifications

Melting Point 242°C
Color Yellow
Quantity 5 g
Formula Weight 214.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Aminodesacetoxycephalosporanic Acid
EINECSNumber (8)-0483
TSCA No