accessibility menu, dialog, popup

UserName

CH3OOH3CH2N

CAS RN 22223-49-0

IUPAC Name: methyl 2-amino-3-methylbenzoate
Synonyms: methyl 2-amino-3-methylbenzoate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: VSFYTPXXMLJNAU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(C)=C1N
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)