accessibility menu, dialog, popup

UserName

CAS RN 22214-42-2

IUPAC Name: 4-(4-hydroxy-3-methoxyphenyl)but-3-en-2-one
Synonyms: 4-(4-hydroxy-3-methoxyphenyl)but-3-en-2-one
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: AFWKBSMFXWNGRE-UHFFFAOYSA-N
SMILES: COC1=C(O)C=CC(C=CC(C)=O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results