accessibility menu, dialog, popup

UserName

CAS RN 2219-30-9

IUPAC Name: hydrogen (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid chloride
Synonyms: hydrogen penicillamine chloride
Molecular Weight (g/mol): 185.67
Molecular Formula: C5H12ClNO2S
InChi Key: CZDHUFYOXKHLME-DFWYDOINSA-N
SMILES: [H+].[Cl-].CC(C)(S)[C@@H](N)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results