missing translation for 'onlineSavingsMsg'
Learn More

D-Pantothenic acid calcium salt, Cell culture reagent grade, MP Biomedicals™

Catalog No. ICN19472180
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19472180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19472180 Supplier MP Biomedicals Inc Supplier No. 0219472180

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2219-30-9
Molecular Formula C5H12ClNO2S
Molecular Weight (g/mol) 185.666
InChI Key CZDHUFYOXKHLME-DFWYDOINSA-N
Synonym d-penicillamine hydrochloride, penicillamine hydrochloride, distamine, unii-5n9jk529zj, usaf el-23, usaf a-1705, d---penicillamine hydrochloride, d-valine, 3-mercapto-, hydrochloride, valine, 3-mercapto-, hydrochloride, d
PubChem CID 92173
IUPAC Name (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid;hydrochloride
SMILES CC(C)(C(C(=O)O)N)S.Cl

Specifications

Quantity 100 g
Formula Weight 238.3
Grade Cell Culture Reagent
Density 0.5g/cm3
Chemical Name or Material D-Pantothenic acid
EINECSNumber 205-278-9
Recommended Storage Store at 2° to 8°C