accessibility menu, dialog, popup

UserName

CH3H3CCH3H2C

CAS RN 2213-32-3

IUPAC Name: 2,4-dimethylpent-1-ene
Synonyms: 2,4-dimethyl-1-pentene
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: LXQPBCHJNIOMQU-UHFFFAOYSA-N
SMILES: CC(C)CC(C)=C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)