missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1-pentene 98.0+%, TCI America™

Catalog No. D12495ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12495ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12495ML Supplier TCI America Supplier No. D12495ML

Chemical Identifiers

CAS 2213-32-3
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00039854
InChI Key LXQPBCHJNIOMQU-UHFFFAOYSA-N
Synonym 2,4-dimethyl-1-pentene, 1-pentene, 2,4-dimethyl, acmc-1ch0z, pentene, 2,4-dimethyl, ch3 2chch2c ch3 =ch2, 1-pentene, 2,4-dimethyl-8ci 9ci
PubChem CID 16657
IUPAC Name 2,4-dimethylpent-1-ene
SMILES CC(C)CC(=C)C

Specifications

Color Colorless
Boiling Point 81°C
Quantity 5 mL
UN Number 3295
Formula Weight 98.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethyl-1-pentene