accessibility menu, dialog, popup

UserName

CAS RN 2210-24-4

IUPAC Name: N-phenylprop-2-enamide
Synonyms: N-phenylprop-2-enamide
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: BPCNEKWROYSOLT-UHFFFAOYSA-N
SMILES: C=CC(=O)NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results