accessibility menu, dialog, popup

UserName

CAS RN 22071-15-4

IUPAC Name: 2-(3-benzoylphenyl)propanoic acid
Synonyms: ketoprofen orudis (TN) profenid orudis KT lertus menamin oruvail ketoprofen preparation dexal oscorel
Molecular Weight (g/mol): 254.29
Molecular Formula: C16H14O3
InChi Key: DKYWVDODHFEZIM-UHFFFAOYNA-N
SMILES: CC(C(O)=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)