missing translation for 'onlineSavingsMsg'
Learn More

Ketoprofen 98.0+%, TCI America™

Catalog No. K003825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K003825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K003825G Supplier TCI America Supplier No. K003825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22071-15-4
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.285
MDL Number MFCD00055790
InChI Key DKYWVDODHFEZIM-UHFFFAOYSA-N
Synonym ketoprofen, 2-3-benzoylphenyl propanoic acid, orudis, m-benzoylhydratropic acid, 2-3-benzoylphenyl propionic acid, ketoprofene, profenid, oruvail, 3-benzoylhydratropic acid, alrheumat
PubChem CID 3825
ChEBI CHEBI:6128
IUPAC Name 2-(3-benzoylphenyl)propanoic acid
SMILES CC(C1=CC=CC(=C1)C(=O)C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 95°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 254.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ketoprofen
RTECSNumber UE7570000
TSCA Yes