missing translation for 'onlineSavingsMsg'
Learn More

Ketoprofen Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019309
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019309 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019309 Supplier Merck Emd Millipore Supplier No. PHR1375500MG

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Ketoprofen is an active pharmaceutical compound used as a non-steroidal anti-inflammatory drug (NSAID). It is essentially a non-narcotic drug extensively utilized for mild to moderate pain relief. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 22071-15-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C16H14O3
MDL Number MFCD00055790
Synonym Ketoprofen; 2-(3-Benzoylphenyl)propionic acid
Type Certified Reference Material
Quantity 500 mg
Formula Weight 254.28
Grade Certified Reference Material