accessibility menu, dialog, popup

UserName

CH3NH2NHO(E/Z)

CAS RN 22059-22-9

IUPAC Name: (Z)-N'-hydroxyethanimidamide
Synonyms: (Z)-N'-hydroxyethanimidamide
Molecular Weight (g/mol): 74.08
Molecular Formula: C2H6N2O
InChi Key: AEXITZJSLGALNH-UHFFFAOYSA-N
SMILES: C\C(N)=N\O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)