accessibility menu, dialog, popup

UserName

HONH2ClFFF

CAS RN 21928-50-7

IUPAC Name: 4-[4-chloro-3-(trifluoromethyl)phenyl]-4-hydroxypiperidin-1-ium
Synonyms: 4-[4-chloro-3-(trifluoromethyl)phenyl]-4-hydroxypiperidin-1-ium
Molecular Weight (g/mol): 280.69
Molecular Formula: C12H14ClF3NO
InChi Key: RALRVIPTUXSBPO-UHFFFAOYSA-O
SMILES: OC1(CC[NH2+]CC1)C1=CC=C(Cl)C(=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results