accessibility menu, dialog, popup

UserName

CAS RN 21834-92-4

IUPAC Name: (2E)-5-methyl-2-phenylhex-2-enal
Synonyms: 5-methyl-2-phenyl-2-hexenal
Molecular Weight (g/mol): 188.27
Molecular Formula: C13H16O
InChi Key: YURDCJXYOLERLO-LCYFTJDESA-N
SMILES: CC(C)C\C=C(\C=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results