missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-phenyl-2-hexenal 90.0+%, TCI America™

Catalog No. M199725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M199725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M199725G Supplier TCI America Supplier No. M199725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21834-92-4
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
MDL Number MFCD00036615
InChI Key YURDCJXYOLERLO-UKTHLTGXSA-N
Synonym 5-methyl-2-phenyl-2-hexenal, 5-methyl-2-phenylhex-2-enal, unii-3et4t99vqa, 3et4t99vqa, z-5-methyl-2-phenylhex-2-enal, 2z-5-methyl-2-phenylhex-2-enal, unii-57uw55ec13 component
PubChem CID 5463934
IUPAC Name (Z)-5-methyl-2-phenylhex-2-enal
SMILES CC(C)CC=C(C=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 100°C
Quantity 25 g
Formula Weight 188.27
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-phenyl-2-hexenal