accessibility menu, dialog, popup

UserName

CAS RN 2168-96-9

H3CSOCH3

CAS RN 2168-96-9

IUPAC Name: 1-(dodecane-1-sulfinyl)dodecane
Synonyms: 1-(dodecane-1-sulfinyl)dodecane
Molecular Weight (g/mol): 386.72
Molecular Formula: C24H50OS
InChi Key: PZKDFFVFMXTDIP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCS(=O)CCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results