missing translation for 'onlineSavingsMsg'
Learn More

Didodecyl Sulfoxide 98.0+%, TCI America™

Catalog No. D28855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28855G 5 g
D288525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28855G Supplier TCI America Supplier No. D28855G

Chemical Identifiers

CAS 2168-96-9
Molecular Formula C24H50OS
Molecular Weight (g/mol) 386.72
MDL Number MFCD00039865
InChI Key PZKDFFVFMXTDIP-UHFFFAOYSA-N
Synonym Dodecyl Sulfoxide
PubChem CID 75109
IUPAC Name 1-(dodecane-1-sulfinyl)dodecane
SMILES CCCCCCCCCCCCS(=O)CCCCCCCCCCCC

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 5 g
Formula Weight 386.72
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Didodecyl Sulfoxide