accessibility menu, dialog, popup

UserName

AlOHOHOH

CAS RN 21645-51-2

IUPAC Name: aluminium(3+) trihydroxide
Synonyms: amphojel|Alu-Cap|AlternaGel
Molecular Weight (g/mol): 78
Molecular Formula: AlH3O3
InChi Key: WNROFYMDJYEPJX-UHFFFAOYSA-K
SMILES: [OH-].[OH-].[OH-].[Al+3]
Grade 
  • (1)
  • (4)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (4)
  • (4)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (7)
  • (1)
  • (4)
  • (7)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (4)
  • (3)
  • (4)