missing translation for 'onlineSavingsMsg'
Learn More

Aluminum hydroxide, extra pure, powder

Catalog No. AC219135000
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219135000 500 g Plastic bottle
AC219130250 25 g Plastic bottle
AC219130020 2 kg Plastic bottle
AC219130100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC219135000 Supplier Thermo Scientific Chemicals Supplier No. 219135000

CAS: 21645-51-2 | AlH3O3 | 78.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21645-51-2
Molecular Formula AlH3O3
Molecular Weight (g/mol) 78.00
MDL Number MFCD00003420
InChI Key WNROFYMDJYEPJX-UHFFFAOYSA-K
IUPAC Name aluminium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Al+3]

Specifications

Color White
pH 8.5 to 10 (5% aq. suspension)
Physical Form Powder
Quantity 500 g
Linear Formula Al(OH)3
Merck Index 15,338
Solubility Information Solubility in water: insoluble
Formula Weight 78
Percent Purity ≥63.5% (Al2O3)
Grade Extra Pure
Loss on Ignition 36.5% max.
Packaging Plastic bottle
Chemical Name or Material Aluminum hydroxide
Show More Show Less
EINECSNumber 244-492-7

RUO – Research Use Only