missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Hydroxide, Powder, 95%, Spectrum™ Chemical

Catalog No. 18608518
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608518 45 kg Poly Pail
18608516 500 g Poly Pail
18608517 2.5 kg Poly Pail
18608515 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608518 Supplier Spectrum Chemical Mfg Cor Supplier No. A109345KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21645-51-2

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 45 kg
Molecular Weight (g/mol) 78.00
Percent Purity 95%
Grade Reagent
Packaging Poly Pail
Chemical Name or Material Aluminum Hydroxide

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.