missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Hydroxide Suspension, For Chloride, Certified, LabChem™

Catalog No. LC108401
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC108401 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC108401 Supplier LabChem Supplier No. LC108401

Chemical Identifiers

CAS 21645-51-2 , 7732-18-5
Molecular Formula AlH3O3
Molecular Weight (g/mol) 78.00
MDL Number MFCD00003420
InChI Key WNROFYMDJYEPJX-UHFFFAOYSA-K
IUPAC Name aluminium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Al+3]

Specifications

Color Undesignated
CAS Max % 97.94, 2.06
Linear Formula Al(OH)3
Solubility Information Soluble in water
Formula Weight 78
Grade Certified
Identification Passes Test
Odor Ammonia
Packaging Poly Bottle
Decomposition Information Aluminum oxides
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Aluminum Hydroxide Suspension, For Chloride
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature