CAS RN 2131-64-8
IUPAC Name:
4-isothiocyanato-N,N-dimethylaniline
Synonyms:
4-isothiocyanato-N,N-dimethylaniline
Molecular Weight (g/mol):
178.25
Molecular Formula:
C9H10N2S
InChi Key:
HRDJPEMAGYHSJR-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C(C=C1)N=C=S
1
–
1
of
1
results
1
Promotions Available
4-Dimethylaminophenyl isothiocyanate, 97%
4-Dimethylaminophenyl isothiocyanate, 97%
CAS: 2131-64-8 Molecular Formula: C9H10N2S Molecular Weight (g/mol): 178.253 MDL Number: MFCD00041093 InChI Key: HRDJPEMAGYHSJR-UHFFFAOYSA-N PubChem CID: 75047 IUPAC Name: 4-isothiocyanato-N,N-dimethylaniline SMILES: CN(C)C1=CC=C(C=C1)N=C=S