accessibility menu, dialog, popup

UserName

CAS RN 212578-38-6

IUPAC Name: 4-methyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid
Synonyms: 4-methyl-2,3-dihydro-1,4-benzoxazine-2-carboxylic acid
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: SSVIRLKDUUXSTR-UHFFFAOYNA-N
SMILES: CN1CC(OC2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results