missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC29801CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC29801CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC29801CB Supplier Thermo Scientific Chemicals Supplier No. CC29801CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 212578-38-6
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD11506351
InChI Key SSVIRLKDUUXSTR-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-2-carboxylic acid, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-2-carboxylic acid, 2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl, 4-methyl-2h,3h-benzo e 1,4-oxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazole-2-carboxylic acid
PubChem CID 10845449
IUPAC Name 4-methyl-2,3-dihydro-1,4-benzoxazine-2-carboxylic acid
SMILES CN1CC(OC2=CC=CC=C21)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 193.2
Percent Purity 97%
Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-2-carboxylic acid
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.