accessibility menu, dialog, popup

UserName

CH3NNHSH2N

CAS RN 21242-21-7

IUPAC Name: (4-methylpyridin-2-yl)thiourea
Synonyms: 4-methylpyridin-2-ylthiourea
Molecular Weight (g/mol): 167.23
Molecular Formula: C7H9N3S
InChi Key: ALLBLAKJCNRLLS-UHFFFAOYSA-N
SMILES: CC1=CC(NC(N)=S)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results