missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Methyl-2-pyridyl)thiourea, 97%

Catalog No. AAH3364806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3364806 5 g
AAH3364803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3364806 Supplier Thermo Scientific Chemicals Supplier No. H3364806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 21242-21-7 | C7H9N3S | 167.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21242-21-7
Molecular Formula C7H9N3S
Molecular Weight (g/mol) 167.23
MDL Number MFCD04117348
InChI Key ALLBLAKJCNRLLS-UHFFFAOYSA-N
Synonym 4-methyl-pyridin-2-yl-thiourea, 4-methylpyridin-2-yl-thiourea, 1-4-methylpyridin-2-yl thiourea, 4-methylpyridin-2-yl thiourea, n-4-methyl-2-pyridyl thiourea, n-4-methylpyridin-2-yl thiourea, thiourea, 4-methyl-2-pyridinyl
PubChem CID 2760059
IUPAC Name (4-methylpyridin-2-yl)thiourea
SMILES CC1=CC(NC(N)=S)=NC=C1

Specifications

Melting Point 216°C to 220°C
Quantity 5 g
Formula Weight 167.23
Percent Purity 97%
Chemical Name or Material N-(4-Methyl-2-pyridyl)thiourea
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only