CAS RN 209986-17-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Thermo Scientific Chemicals Compound E
CAS: 209986-17-4 Molecular Formula: C27H24F2N4O3 Synonym: γ-Secretase Inhibitor XXI;
2
R&D Systems™ Compound E
A Tocris product, part of R&D Systems. γ-secretase inhibitor; induces neuronal differentiation