missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Compound E

Catalog No. AAJ65131EXD
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65131EXD 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65131EXD Supplier Thermo Scientific Chemicals Supplier No. J65131EXD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 209986-17-4 | C27H24F2N4O3

A cell-permeable, potent, selective, peptidomimetic, non-transition-state analog inhibitor of --secretase and Notch processing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 209986-17-4
Molecular Formula C27H24F2N4O3
Synonym γ-Secretase Inhibitor XXI;

Specifications

Quantity 0.5 mg
Description Off-white
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable, potent, selective, peptidomimetic, non-transition-state analog inhibitor of →-secretase and Notch processing
Formula Weight 490.50
Sensitivity Light Sensitive
Form Solid

RUO – Research Use Only