accessibility menu, dialog, popup

UserName

CAS RN 20967-96-8

ON

CAS RN 20967-96-8

IUPAC Name: 2-[3-(benzyloxy)phenyl]acetonitrile
Synonyms: 2-[3-(benzyloxy)phenyl]acetonitrile
Molecular Weight (g/mol): 223.28
Molecular Formula: C15H13NO
InChi Key: CKZFVIPFANUBDW-UHFFFAOYSA-N
SMILES: N#CCC1=CC(OCC2=CC=CC=C2)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results