missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxyphenylacetonitrile 98.0+%, TCI America™

Catalog No. B42291G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B42291G Supplier TCI America Supplier No. B42291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20967-96-8
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.28
MDL Number MFCD00016396
InChI Key CKZFVIPFANUBDW-UHFFFAOYSA-N
Synonym 3-benzyloxyphenylacetonitrile, 2-3-benzyloxy phenyl acetonitrile, 3-benzyloxybenzyl cyanide, 2-3-phenylmethoxyphenyl acetonitrile, 2-3-phenylmethoxy phenyl ethanenitrile, maybridge3_000453, 3-benzyloxyphenyl acetonitrile, 3-benzyloxy-phenyl acetonitrile, 3-benzyloxyphenyl-acetonitrile
PubChem CID 519831
IUPAC Name 2-[3-(benzyloxy)phenyl]acetonitrile
SMILES N#CCC1=CC(OCC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 175°C
Quantity 1 g
Formula Weight 223.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzyloxyphenylacetonitrile