accessibility menu, dialog, popup

UserName

ONN

CAS RN 20917-49-1

IUPAC Name: 1-nitrosoazocane
Synonyms: N-nitroso-heptamethyleneimine
Molecular Weight (g/mol): 142.2
Molecular Formula: C7H14N2O
InChi Key: RCFKQVLHWYOSFF-UHFFFAOYSA-N
SMILES: O=NN1CCCCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results