accessibility menu, dialog, popup

UserName

CAS RN 2081-44-9

OHO

CAS RN 2081-44-9

IUPAC Name: oxan-4-ol
Synonyms: oxan-4-ol
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: LMYJGUNNJIDROI-UHFFFAOYSA-N
SMILES: OC1CCOCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results